1-Phenyl-1-decanon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Decyl-phenylketon

311286-2G
SIAL311286-2GEA 69.9 EUR
SIAL311286-2G
1-Phenyl-1-decanon, Sigma-Aldrich®
1-Phenyl-1-decanon
Formel: C₆H₅C(O)(CH₂)₈CH₃
Molecular Weight: 232,37 g/mol
Siedepunkt: 120…128 °C (0,5 mmHg)
Schmelzpunkt: 35 °C
MDL: MFCD00009582
CAS-Nummer: 6048-82-4
EINECS: 227-946-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=39f810acd80d51fb36e51c955463f46d95781d5d&vid=bcade9ec37705caa44872db67f6a31b6b848c12c&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR