Dexamethason, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 9-Fluor-11β,17α,21-trihydroxy-16α-methyl-pregna-1,4-dien-3,20-dion, Dexamethasonum

M-Clarity™ Qualitätsstufe = MQ100

31375-500MG
SIAL31375-500MGEA 197 EUR
SIAL31375-500MG
Dexamethason, Sigma-Aldrich®
Dexamethason
Formel: C₂₂H₂₉FO₅
Molecular Weight: 392.47 g/mol
Schmelzpunkt: 268…271 °C
MDL: MFCD00064136
CAS-Nummer: 50-02-2
EINECS: 200-003-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR