Dextran, M.W. ~ 60,000 (aus Leuconostoc mesenteroides), BioChemika®, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ200
31397-100G
SIAL31397-100GEA
279
EUR
SIAL31397-100G
Dextran, M.W. ~ 60,000 (aus Leuconostoc mesenteroides), BioChemika®, Sigma-Aldrich®
Dextran
Formel:
(C₆H₁₀O₅)n |
MDL:
MFCD00130935 CAS-Nummer: 9004-54-0 EINECS: 232-677-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...