3-Brombenzylamin Hydrochlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (3-Bromophenyl)methanamine hydrochloride

M-Clarity™ Qualitätsstufe = MQ100

316288-25G
SIAL316288-25GEA 130 EUR
SIAL316288-25G
3-Brombenzylamin Hydrochlorid, Sigma-Aldrich®
3-Brombenzylamin Hydrochlorid
Formel: C₇H₉BrClN
Molecular Weight: 222,52 g/mol
MDL: MFCD00012856
CAS-Nummer: 39959-54-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR