4-Isopropyl-m-kresol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

316431-100G
SIAL316431-100GEA 124 EUR
SIAL316431-100G
4-Isopropyl-m-kresol, Sigma-Aldrich®
4-Isopropyl-m-kresol
Formel: C₁₀H₁₄O
MDL: MFCD00010704
CAS-Nummer: 3228-02-2
EINECS: 221-761-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR