4-Hydroxy-3-methylbenzaldehyd, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

316911-1G
SIAL316911-1GEA 45.7 EUR
SIAL316911-1G
4-Hydroxy-3-methylbenzaldehyd, Sigma-Aldrich®
4-Hydroxy-3-methylbenzaldehyde
Formel: C₈H₈O₂
Molecular Weight: 136,1479 g/mol
MDL: MFCD00012360
CAS-Nummer: 15174-69-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR