(R)-(+)-1-Phenyl-1-butanol, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
(R)-1-Phenylbutan-1-ol
317314-1G
SIAL317314-1GEA
142
EUR
SIAL317314-1G
(R)-(+)-1-Phenyl-1-butanol, Sigma-Aldrich®
(R)-(+)-1-Phenyl-1-butanol
Formel:
C₁₀H₁₄O Molecular Weight: 150,22 g/mol Siedepunkt: 232 °C (1013 hPa) Schmelzpunkt: 31…42,5 °C Dichte: 0,978 g/cm³ (20 °C) |
MDL:
MFCD00004575 CAS-Nummer: 22144-60-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...