Allylphenylsulfon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (Allylsulfonyl)benzol

M-Clarity™ Qualitätsstufe = MQ200

317713-25G
SIAL317713-25GEA 125 EUR
SIAL317713-25G
Allylphenylsulfon, Sigma-Aldrich®
Allylphenylsulfon
Formel: C₉H₁₀O₂S
Molecular Weight: 182,24 g/mol
MDL: MFCD00014739
CAS-Nummer: 16212-05-8
EINECS: 240-338-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR