Natriumphenoxid Trihydrat, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Sodium phenolate trihydrate
M-Clarity™ Qualitätsstufe = MQ100
318191-100G
318191-5G
SIAL318191-100GEA
59.7
EUR
SIAL318191-100G
SIAL318191-5G
Natriumphenoxid Trihydrat, Sigma-Aldrich®
Natriumphenoxid Trihydrat
Formel:
C₆H₅NaO·3H₂O |
MDL:
MFCD00149636 CAS-Nummer: 156150-40-2 EINECS: 205-347-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...