Ferroin (1,10-Phenanthrolin-Eisen(II)sulfat-Komplex) 0,1% in wässriger Lösung Indikator
Lieferant: Merck
Synonyme:
Tris(1,10-phenanthrolin-N1,N10)eisensulfat
318922-100ML
SIAL318922-100MLEA
60.8
EUR
SIAL318922-100ML
Ferroin (1,10-Phenanthrolin-Eisen(II)sulfat-Komplex) 0,1% in wässriger Lösung Indikator
Ferroin (1,10-Phenanthrolin-Eisen(II)sulfat-Komplex)
Formel:
C₃₆H₂₄FeN₆SO₄ Molecular Weight: 692,54 g/mol |
MDL:
MFCD00036428 CAS-Nummer: 14634-91-4 EINECS: 238-676-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...