Cyclopentanthiol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Mercaptocyclopentan, Cyclopentylmercaptan

M-Clarity™ Qualitätsstufe = MQ100

319708-5G
SIAL319708-5GEA 41.2 EUR
SIAL319708-5G
Cyclopentanthiol, Sigma-Aldrich®
Cyclopentanthiol
Formel: C₅H₁₀S
Molecular Weight: 102.2 g/mol
Siedepunkt: 131…132 °C (1013 hPa)
Dichte: 0.957 g/cm³ (20 °C)
MDL: MFCD00001369
CAS-Nummer: 1679-07-8
EINECS: 216-841-3
UN: 3336
ADR: 3 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR