3,5-Dichlor-1,2-dinitrobenzol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1,5-Dichloro-2,3-dinitrobenzene

M-Clarity™ Qualitätsstufe = MQ100

319759-5G
SIAL319759-5GEA 183 EUR
SIAL319759-5G
3,5-Dichlor-1,2-dinitrobenzol, Sigma-Aldrich®
3,5-Dichlor-1,2-dinitrobenzol
Formel: C₆H₂Cl₂N₂O₄
Molecular Weight: 237 g/mol
CAS-Nummer: 28689-08-9
EINECS: 249-157-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR