3-Fluorsalicylaldehyd, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-Fluoro-2-hydroxybenzaldehyde

M-Clarity™ Qualitätsstufe = MQ200

319805-1G
SIAL319805-1GEA 38.9 EUR
SIAL319805-1G
3-Fluorsalicylaldehyd, Sigma-Aldrich®
3-Fluorsalicylaldehyd
Formel: C₇H₅FO₂
Molecular Weight: 140,12 g/mol
MDL: MFCD00003319
CAS-Nummer: 394-50-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR