Acetophenon-[Ring-D5], Sigma-Aldrich®

Lieferant: Merck

Synonyme: Phenyl-methylketon-2',3',4',5',6'-D5, Methyl-phenylketon-2',3',4',5',6'-D5, 1-Phenyl-1-ethanon-2',3',4',5',6'-D5, Ethanoylbenzol-2',3',4',5',6'-D5

M-Clarity™ Qualitätsstufe = MQ200

319856-5G
SIAL319856-5GEA 489 EUR
SIAL319856-5G
Acetophenon-[Ring-D5], Sigma-Aldrich®
Acetophenon-[Ring-D5]
Formel: C₈H₃D₅O
Molecular Weight: 125,11 g/mol
CAS-Nummer: 28077-64-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR