Aceton Reagens ≥99,5% ACS

Lieferant: Merck
Gefahr


M-Clarity™ Qualitätsstufe = MQ200

320110-1L
SIAL320110-1LEA 69.8 EUR
SIAL320110-1L
Aceton Reagens ≥99,5% ACS
Aceton

Acetone is a polar, aprotic organic solvent. It is a colourless and highly volatile liquid. It is commonly used in organic syntheses as solvent and/or building block, in pharmaceutical and personal care industries. This grade meets ACS requirements, making it ideal for a wide range of analytical applications.


  • Used as solvent and/or building block in organic syntheses, such as SN2 reactions and Jones oxidation
  • Mobile phase in Thin layer chromatography (TLC)
  • Used as solvent in chemical molecules delivery systems in pharmaceutical and cosmetic industries
  • Cleaning proposes
  • Acetone's luminesence intensity is dependent upon the solution components
  • The absorption of UV light by acetone, results in its photolysis and the production of radials


ACS solvents meet or exceed the high standards of the American Chemical Society (ACS), with test specifications that are specialised to every compound. According to the American Chemical Society, ACS reagent grade implies that it is a substance of sufficient purity to be used in most chemical analyses or reactions.

Formel: CH₃COCH₃
Siedepunkt: 56 °C/760 mmHg (Lit.)
Schmelzpunkt: –94 °C (Lit.)
Dichte: 0,791 g/ml (25 °C) (Lit.)
Flammpunkt: −17,0 °C
MDL: MFCD00008765
CAS-Nummer: 67-64-1
UN: 1090
ADR: 3,II

Bestellen

Spezifikation Testergebnisse

Appearance (Colour) Colourless
Appearance (Form) Liquid
Infrared Spectrum Conforms to Structure
Colour Test ≤10 APHA
Purity (GC)
Methanol Content
≤0.05%
GC Identified Peak
Isopropyl Alcohol
≤0.05%
Corrected for Water Content
Purity by GC
≥99.50%
Residue on Evaporation ≤0.001%
Water (by Karl Fischer) ≤0.5%
Titrable Acid (meq/g) < 0.0003 ≤0.0003
Titrable Base (meq/g) ≤0.0006
Aldehyde (as Formaldehyde) ≤0.002%
Substances reducing MnO4 Pass
Solubility Pass
Meets ACS Requirements Pass
Recommended Retest Period Current ACS Specification
2 Years


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR