Ethyl-4-oxocyclohexancarboxylat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Ethyl 4-oxocyclohexanecarboxylate, 4-(Ethoxycarbonyl)-1-cyclohexanone

320625-1G
SIAL320625-1GEA 57.1 EUR
SIAL320625-1G
Ethyl-4-oxocyclohexancarboxylat, Sigma-Aldrich®
Ethyl-4-oxocyclohexancarboxylat
Formel: C₉H₁₄O₃
Molecular Weight: 170,21 g/mol
MDL: MFCD00013285
CAS-Nummer: 17159-79-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR