1,3-Dimethoxy-5-fluorbenzol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 5-Fluorresorcindimethylether

M-Clarity™ Qualitätsstufe = MQ100

322342-5G
SIAL322342-5GEA 81.1 EUR
SIAL322342-5G
1,3-Dimethoxy-5-fluorbenzol, Sigma-Aldrich®
1,3-Dimethoxy-5-fluorbenzol
Formel: C₈H₉FO₂
Molecular Weight: 156,16 g/mol
MDL: MFCD00012445
CAS-Nummer: 52189-63-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR