4-Brom-1,8-naphthalsäureanhydrid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 6-Bromobenzo[de]isochromene-1,3-dione

M-Clarity™ Qualitätsstufe = MQ100

324248-5G
SIAL324248-5GEA 60.3 EUR
SIAL324248-5G
4-Brom-1,8-naphthalsäureanhydrid, Sigma-Aldrich®
4-Brom-1,8-naphthalsäureanhydrid
Formel: C₁₂H₅BrO₃
Molecular Weight: 277,0703 g/mol
MDL: MFCD00006927
CAS-Nummer: 81-86-7
EINECS: 201-382-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR