2,5-Dimethylphenylhydraziniumchlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-(2,5-Dimethylphenyl)hydrazin Hydrochlorid, 2,5-Dimethylphenylhydrazinhydrochlorid, 2,5-Dimethylphenylhydrazin Hydrochlorid

M-Clarity™ Qualitätsstufe = MQ200

324299-25G
SIAL324299-25GEA 78.1 EUR
SIAL324299-25G
2,5-Dimethylphenylhydraziniumchlorid, Sigma-Aldrich®
2,5-Dimethylphenylhydraziniumchlorid
Formel: C₈H₁₃ClN₂
Molecular Weight: 172,66 g/mol
MDL: MFCD00013382
CAS-Nummer: 56737-78-1
UN: 2811
ADR: 6.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR