4-Cyan-2-nitroanilin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Cyano-2-nitroaniline

M-Clarity™ Qualitätsstufe = MQ100

324698-5G
SIAL324698-5GEA 51.3 EUR
SIAL324698-5G
4-Cyan-2-nitroanilin, Sigma-Aldrich®
4-Cyan-2-nitroanilin
Formel: C₇H₅N₃O₂
Molecular Weight: 163,14 g/mol
MDL: MFCD00013373
CAS-Nummer: 6393-40-4
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR