4-Fluor-2-nitroanisol, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: 4-Fluoro-1-methoxy-2-nitrobenzene

M-Clarity™ Qualitätsstufe = MQ200

324760-5G
SIAL324760-5GEA 56.3 EUR
SIAL324760-5G
4-Fluor-2-nitroanisol, Sigma-Aldrich®
4-Fluor-2-nitroanisol
Formel: C₇H₆FNO₃
Molecular Weight: 171,13 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00013375
CAS-Nummer: 445-83-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR