Ethyl-2-methylnicotinat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Ethyl 2-methylnicotinate

M-Clarity™ Qualitätsstufe = MQ100

325201-5G
SIAL325201-5GEA 77.4 EUR
SIAL325201-5G
Ethyl-2-methylnicotinat, Sigma-Aldrich®
Ethyl-2-methylnicotinat
Formel: C₉H₁₁NO₂
Molecular Weight: 165,19 g/mol
MDL: MFCD00006336
CAS-Nummer: 1721-26-2
EINECS: 217-013-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR