Cer(III)oxalat Hydrat, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: Cerium(III) oxalate hydrate

M-Clarity™ Qualitätsstufe = MQ200

325511-250G
SIAL325511-250GEA 99.7 EUR
SIAL325511-250G
Cer(III)oxalat Hydrat, Sigma-Aldrich®
Cer(III)oxalat Hydrat
Formel: C₆Ce₂O₁₂·H₂O
Molecular Weight: 544,29 g/mol (anhy)
MDL: MFCD00150139
CAS-Nummer: 15750-47-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR