Lanthan(III)carbonat Hydrat, Sigma-Aldrich®

Lieferant: Merck

325767-100G
SIAL325767-100GEA 80 EUR
SIAL325767-100G
Lanthan(III)carbonat Hydrat, Sigma-Aldrich®
Lanthan(III)carbonat Hydrat
Formel: La₂(CO₃)₃.xH₂O
Molecular Weight: 457,85 g/mol (anhy)
MDL: MFCD00011069
CAS-Nummer: 54451-24-0
EINECS: 209-599-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR