Neodym(III)sulfat Hydrat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

325813-50G
SIAL325813-50GEA 201 EUR
SIAL325813-50G
Neodym(III)sulfat Hydrat, Sigma-Aldrich®
Neodym(III)sulfat Hydrat
Formel: Nd₂S₃O₁₂·H₂O
CAS-Nummer: 101509-27-7
EINECS: 233-262-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR