Samarium(III)sulfat Octahydrat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

325880-25G
SIAL325880-25GEA 93.6 EUR
SIAL325880-25G
Samarium(III)sulfat Octahydrat, Sigma-Aldrich®
Samarium(III)sulfat Octahydrat
Formel: O₁₂S₃Sm₂·8H₂O
Molecular Weight: 733,03 g/mol
MDL: MFCD00011236
CAS-Nummer: 13465-58-2
EINECS: 237-211-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR