Terbium(III)sulfat Octahydrat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Diterbiumtrisulfat Octahydrat

325953-5G
SIAL325953-5GEA 46.4 EUR
SIAL325953-5G
Terbium(III)sulfat Octahydrat, Sigma-Aldrich®
Terbium(III)sulfat Octahydrat
Formel: O₁₄S₃Tb₂·8H₂O
MDL: MFCD00149872
CAS-Nummer: 13842-67-6
EINECS: 237-213-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR