Dithuliumtricarbonat Hydrat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

325988-1G
SIAL325988-1GEA 156 EUR
SIAL325988-1G
Dithuliumtricarbonat Hydrat, Sigma-Aldrich®
Dithuliumtricarbonat Hydrat
Formel: C₃O₉Tm₂·H₂O
CAS-Nummer: 87198-17-2
EINECS: 227-887-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR