Tetrachlorbis(tetrahydrofuran)niob(IV), Sigma-Aldrich®

Lieferant: Merck

326364-5G
SIAL326364-5GEA 97.8 EUR
SIAL326364-5G
Tetrachlorbis(tetrahydrofuran)niob(IV), Sigma-Aldrich®
Tetrachlorbis(tetrahydrofuran)niob(IV)
Formel: C₄H₈Cl₄NbO
MDL: MFCD00013405
CAS-Nummer: 61247-57-2
UN: 3261
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR