Gold, Pulver, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
326585-500MG
326585-5G
SIAL326585-500MGEA
344
EUR
SIAL326585-500MG
SIAL326585-5G
Gold, Pulver, Sigma-Aldrich®
Gold
Formel:
Au Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003436 CAS-Nummer: 7440-57-5 EINECS: 231-165-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...