3,3'-Difluorbenzophenon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Bis(3-fluorphenyl)keton

M-Clarity™ Qualitätsstufe = MQ100

327174-5G
SIAL327174-5GEA 148 EUR
SIAL327174-5G
3,3'-Difluorbenzophenon, Sigma-Aldrich®
3,3'-Difluorbenzophenon
Formel: C₁₃H₈F₂O
Molecular Weight: 218,2 g/mol
MDL: MFCD00000337
CAS-Nummer: 345-70-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR