Pentacarbonylrhenium(I)bromid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

327638-1G
SIAL327638-1GEA 186 EUR
SIAL327638-1G
Pentacarbonylrhenium(I)bromid, Sigma-Aldrich®
Pentacarbonylrhenium(I)bromid
Formel: C₅BrO₅Re
MDL: MFCD00013295
CAS-Nummer: 14220-21-4
EINECS: 238-084-8
UN: 3466
ADR: 6.1,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR