4-Brom-2,6-difluoranilin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

327867-5G
SIAL327867-5GEA 89.8 EUR
SIAL327867-5G
4-Brom-2,6-difluoranilin, Sigma-Aldrich®
4-Brom-2,6-difluoranilin
Formel: C₆H₄BrF₂N
Molecular Weight: 208,01 g/mol
MDL: MFCD00013389
CAS-Nummer: 67567-26-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR