2-Methylhippursäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

328006-1G 328006-5G
SIAL328006-1GEA 43.9 EUR
SIAL328006-1G SIAL328006-5G
2-Methylhippursäure, Sigma-Aldrich®
2-Methylhippursäure
Formel: C₁₀H₁₁NO₃
MDL: MFCD00050991
CAS-Nummer: 42013-20-7
EINECS: 255-622-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR