4,4'-Dibrombenzil, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: Bis(4-bromophenyl)ethanedione

M-Clarity™ Qualitätsstufe = MQ100

328057-10G
SIAL328057-10GEA 68 EUR
SIAL328057-10G
4,4'-Dibrombenzil, Sigma-Aldrich®
4,4'-Dibrombenzil
Formel: C₁₄H₈Br₂O₂
Molecular Weight: 368,02 g/mol
MDL: MFCD00000104
CAS-Nummer: 35578-47-3
EINECS: 252-628-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR