Tetramethylammoniumhydroxid 10% aqueous solution, Sigma-Aldrich®

Lieferant: Merck
Gefahr


MQ100

328251-1L 328251-250ML
SIAL328251-1LEA 164 EUR
SIAL328251-1L SIAL328251-250ML
Tetramethylammoniumhydroxid 10% aqueous solution, Sigma-Aldrich®
Tetramethylammonium hydroxide
Formel: C₄H₁₃NO
Molecular Weight: 91.15 g/mol
Dichte: 1.02 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00008280
CAS-Nummer: 75-59-2
EINECS: 200-882-9
UN: 1835
ADR: 8 II
Merck Index: 14,09224

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR