(R)-(-)-1-Indanol, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: (R)-(-)-1-Hydroxyindan

M-Clarity™ Qualitätsstufe = MQ200

328413-1G
SIAL328413-1GEA 318 EUR
SIAL328413-1G
(R)-(-)-1-Indanol, Sigma-Aldrich®
(R)-(-)-1-Indanol
Formel: C₉H₁₀O
Molecular Weight: 134,18 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00064164
CAS-Nummer: 697-64-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR