4-Cyan-4'-pentylbiphenyl, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4'-Pentyl-4-biphenylcarbonitrile, 4'-Pentyl-[1,1'-biphenyl]-4-carbonitrile

M-Clarity™ Qualitätsstufe = MQ100

328510-1G
SIAL328510-1GEA 208 EUR
SIAL328510-1G
4-Cyan-4'-pentylbiphenyl, Sigma-Aldrich®
4-Cyan-4'-pentylbiphenyl
Formel: C₁₈H₁₉N
Molecular Weight: 249,36 g/mol
MDL: MFCD00036350
CAS-Nummer: 40817-08-1
EINECS: 255-093-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR