4-Cyan-4'-pentyloxybiphenyl, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Amyloxy-4'-cyanobiphenyl, 4-Cyano-4'-n-pentyloxybiphenyl, 5OCB, 4'-Amyloxy-4-biphenylcarbonitrile, 4'-Pentyloxy-4-biphenylcarbonitrile, 4'-Pentyloxy-[1,1'-biphenyl]-4-carbonitrile

M-Clarity™ Qualitätsstufe = MQ200

328529-1G
SIAL328529-1GEA 0 EUR
SIAL328529-1G
4-Cyan-4'-pentyloxybiphenyl, Sigma-Aldrich®
4-Cyan-4'-pentyloxybiphenyl
Formel: C₁₈H₁₉NO
Molecular Weight: 265,35 g/mol
MDL: MFCD00074879
CAS-Nummer: 52364-71-3
EINECS: 257-875-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR