2-Nitro-α,α,α-trifluor-p-toluylsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Nitro-4-(trifluormethyl)benzoesäure

M-Clarity™ Qualitätsstufe = MQ100

328901-1G
SIAL328901-1GEA 27 EUR
SIAL328901-1G
2-Nitro-α,α,α-trifluor-p-toluylsäure, Sigma-Aldrich®
2-Nitro-α,α,α-trifluor-p-toluylsäure
Formel: C₈H₄F₃NO₄
Molecular Weight: 235,12 g/mol
MDL: MFCD00007142
CAS-Nummer: 320-94-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR