2,3,5,6-Tetrafluorbenzaldehyd, Sigma-Aldrich®

Lieferant: Merck
Achtung


M-Clarity™ Qualitätsstufe = MQ100

328936-1G
SIAL328936-1GEA 41.5 EUR
SIAL328936-1G
2,3,5,6-Tetrafluorbenzaldehyd, Sigma-Aldrich®
2,3,5,6-Tetrafluorobenzaldehyde
Formel: C₇H₂F₄O
Molecular Weight: 178,09 g/mol
MDL: MFCD00167387
CAS-Nummer: 19842-76-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR