Geranylbromid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

329118-25G 329118-5G
SIAL329118-25GEA 218 EUR
SIAL329118-25G SIAL329118-5G
Geranylbromid, Sigma-Aldrich®
Geranylbromid
Formel: C₁₀H₁₇Br
MDL: MFCD00000243
CAS-Nummer: 6138-90-5
EINECS: 228-123-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR