4,5-Dimethoxy-2-nitrobenzoesäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

329231-100G
SIAL329231-100GEA 188 EUR
SIAL329231-100G
4,5-Dimethoxy-2-nitrobenzoesäure, Sigma-Aldrich®
4,5-Dimethoxy-2-nitrobenzoesäure
Formel: C₉H₉NO₆
Molecular Weight: 227.17 g/mol
MDL: MFCD00014697
CAS-Nummer: 4998-07-6
EINECS: 225-657-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR