1,4,7-Triazacyclononan Trihydrochlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1,4,7-Triazonane trihydrochloride

329495-1G
SIAL329495-1GEA 170 EUR
SIAL329495-1G
1,4,7-Triazacyclononan Trihydrochlorid, Sigma-Aldrich®
1,4,7-Triazacyclononan Trihydrochlorid
Formel: C₆H₁₈Cl₃N₃
Molecular Weight: 238,5900 g/mol
MDL: MFCD00074887
CAS-Nummer: 58966-93-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR