Zinc undecylenate 98% (durch komplexometrische Analyse), Sigma-Aldrich®

Lieferant: Merck

Synonyme: 10-Undecenoic Acid Zinc

M-Clarity™ Qualitätsstufe = MQ100

329584-500G
SIAL329584-500GEA 295 EUR
SIAL329584-500G
Zinc undecylenate 98% (durch komplexometrische Analyse), Sigma-Aldrich®
Zinc undecylenate
Formel: C₂₂H₃₈O₄Zn
Molecular Weight: 431,9437 g/mol
Schmelzpunkt: 118,..121 °C (Lit.)
MDL: MFCD00067246
CAS-Nummer: 557-08-4
EINECS: 209-155-0

Bestellen

Spezifikation Testergebnisse

Appearance (Colour) White
Appearance (Form) Powder
Infrared Spectrum Conforms to Structure
Complexiometric EDTA 97.5 - 102.5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR