3,4-(Methylendioxy)phenylessigsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (1,3-Benzodioxol-5-yl)acetic acid

329673-5G
SIAL329673-5GEA 147 EUR
SIAL329673-5G
3,4-(Methylendioxy)phenylessigsäure, Sigma-Aldrich®
3,4-(Methylendioxy)phenylessigsäure
Formel: C₉H₈O₄
Molecular Weight: 180,16 g/mol
MDL: MFCD00014576
CAS-Nummer: 2861-28-1
EINECS: 220-679-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR