2-Nitro-4-(trifluormethyl)benzonitril, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

329835-5G
SIAL329835-5GEA 65.6 EUR
SIAL329835-5G
2-Nitro-4-(trifluormethyl)benzonitril, Sigma-Aldrich®
2-Nitro-4-(trifluormethyl)benzonitril
Formel: C₈H₃F₃N₂O₂
MDL: MFCD00014684
CAS-Nummer: 778-94-9
EINECS: 212-298-1
UN: 3439
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR