2-Brom-3-nitrobenzoesäure, Sigma-Aldrich®

Lieferant: Merck
Achtung

330191-1G
SIAL330191-1GEA 75.2 EUR
SIAL330191-1G
2-Brom-3-nitrobenzoesäure, Sigma-Aldrich®
2-Bromo-3-nitrobenzoic acid
Formel: C₇H₄BrNO₄
Molecular Weight: 246,02 g/mol
MDL: MFCD00074899
CAS-Nummer: 573-54-6
EINECS: 209-356-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR