4'-Hydroxy-3'-nitroacetophenon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-(4-Hydroxy-3-nitrophenyl)-1-ethanone

M-Clarity™ Qualitätsstufe = MQ100

330256-1G
SIAL330256-1GEA 41.3 EUR
SIAL330256-1G
4'-Hydroxy-3'-nitroacetophenon, Sigma-Aldrich®
4'-Hydroxy-3'-nitroacetophenon
Formel: C₈H₇NO₄
Molecular Weight: 181,15 g/mol
MDL: MFCD00017002
CAS-Nummer: 6322-56-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR