3,5-Dimethoxybenzylchlorid, Sigma-Aldrich®

Lieferant: Merck

330264-5G
SIAL330264-5GEA 287 EUR
SIAL330264-5G
3,5-Dimethoxybenzylchlorid, Sigma-Aldrich®
3,5-Dimethoxybenzylchlorid
Formel: C₉H₁₁ClO₂
Molecular Weight: 186,64 g/mol
MDL: MFCD00074902
CAS-Nummer: 6652-32-0
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR